Thermo Scientific Chemicals

Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals

Catalog number: 018595.30
250 g, Each
Thermo Scientific Chemicals

Di-n-butyltin diacetate, 95%, Thermo Scientific Chemicals

Catalog number: 018595.30
250 g, Each
Quantity
Catalog number: 018595.30
also known as 018595-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1067-33-0
Molecular Formula
C12H26O4Sn
InChI Key
LDZDQDDROJXUBO-UHFFFAOYSA-N
SMILES
CC(O)=O.CC(O)=O.CCCC[SnH2]CCCC
Molecular Weight (g/mol)
353.05
Assay (Titration)
32.2 to 34.2 wt% total Sn
Appearance (Color)
Colorless to pale yellow to pale brown
Form
Liquid
Assay (unspecified)
33.6 to 35.6 wt% acetic acid

Description

As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions.Di-n-butyltin diacetate is used as a stabilizer for polyvinyl chloride. It acts as a catalyst for the preparation of silicone and urethane foams. Further, it is used as a curing agent for silicone elastomers. In addition to this, it is used as a precursor for thin-film tin(IV) oxide gas sensing materials by laser-assisted chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text