Thermo Scientific Chemicals

Silver methanesulfonate, Thermo Scientific Chemicals

Catalog number: 018641.18
50 g, Each
Thermo Scientific Chemicals

Silver methanesulfonate, Thermo Scientific Chemicals

Catalog number: 018641.18
50 g, Each
Quantity
Catalog number: 018641.18
also known as 018641-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2386-52-9
IUPAC Name
methanesulfonic acid silver
Molecular Formula
CH4AgO3S
InChI Key
GTBQRHOYAUGRPV-UHFFFAOYSA-N
SMILES
[Ag].CS(O)(=O)=O
Assay (unspecified)
50% Ag min

Description

Silver methanesulfonate is used as a catalyst for heterocyclization reactions. Further, it is used in the synthesis of unsaturated ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text