Thermo Scientific Chemicals

Holmium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 018681.06
5 g, Each
Thermo Scientific Chemicals

Holmium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 018681.06
5 g, Each
Quantity
Catalog number: 018681.06
also known as 018681-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10138-62-2
IUPAC Name
holmium(3+) trichloride
Molecular Formula
Cl3Ho
InChI Key
PYOOBRULIYNHJR-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ho+3]
Total Metal Impurities
0.1% max.

Description

Holmium(III) chloride has many application in the field of nuclear industry, lasers research applications. It is also employed in calibration of optical spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text