Holmium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Holmium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 018681.14
also known as 018681-14
Price (USD)/ Each
511.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
511.00
Add to cart
Holmium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number018681.14
Price (USD)/ Each
511.00
-
Add to cart
Chemical Identifiers
CAS10138-62-2
IUPAC Nameholmium(3+) trichloride
Molecular FormulaCl3Ho
InChI KeyPYOOBRULIYNHJR-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Holmium(III) chloride has many application in the field of nuclear industry, lasers research applications. It is also employed in calibration of optical spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only