Thermo Scientific Chemicals

Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 018685.03
1 g, Each
Thermo Scientific Chemicals

Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 018685.03
1 g, Each
Quantity
Catalog number: 018685.03
also known as 018685-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10099-66-8
IUPAC Name
lutetium(3+) trichloride
Molecular Formula
Cl3Lu
InChI Key
AEDROEGYZIARPU-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Lu+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Lutetium(III) chloride, anhydrous is used as a sputtering target in the preparation of bubble memory device. It is widely used in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text