Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 018685.06
also known as 018685-06
Price (USD)/ Each
660.65
Online exclusive
734.00 
Save 73.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
660.65
Online exclusive
734.00 
Save 73.35 (10%)
Add to cart
Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Catalog number018685.06
Price (USD)/ Each
660.65
Online exclusive
734.00 
Save 73.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10099-66-8
IUPAC Namelutetium(3+) trichloride
Molecular FormulaCl3Lu
InChI KeyAEDROEGYZIARPU-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Lu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Lutetium(III) chloride, anhydrous is used as a sputtering target in the preparation of bubble memory device. It is widely used in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only