Thermo Scientific Chemicals
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate, Thermo Scientific Chemicals
Catalog number: 018809.02
0.5 g, Each
Thermo Scientific Chemicals
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate, Thermo Scientific Chemicals
Catalog number: 018809.02
0.5 g, Each
Quantity
Catalog number: 018809.02
also known as 018809-02
Price (USD)
Have Questions?
Description
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is used as a catalyst for isomerization of allylic ethers of glycerol and glucose to the corresponding 1-propenyl ethers. It is also used as a catalyst in hydrogenation reactions.