Thermo Scientific Chemicals

Nickel(II) 2,4-pentanedionate, 95%, Thermo Scientific Chemicals

Catalog number: 018811.36
500 g, Each
Thermo Scientific Chemicals

Nickel(II) 2,4-pentanedionate, 95%, Thermo Scientific Chemicals

Catalog number: 018811.36
500 g, Each
Quantity
Catalog number: 018811.36
also known as 018811-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3264-82-2
IUPAC Name
nickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C10H14NiO4
InChI Key
BMGNSKKZFQMGDH-FDGPNNRMSA-L
SMILES
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Appearance (Color)
Green
Form
Powder or crystalline powder or crystals
Assay from Supplier's CofA
Nickel, Ni : 21.5-24.2%
Assay from Supplier's CofA
≥94.0 to ≤106.0% (Titration)

Description

Catalyst and U.V. stabilizer.Nickel(II) 2,4-pentanedionate is used as a precursor to nickel bis(cyclooctadiene) catalyst. It is also used in the deposition of nickel(II) oxide thin film by sol-gel techniques on conductive glass substrates. Further, it is used in organic synthesis to produce organometals. It is associated with dimethylgold(III) acetylacetonate is used in gold on nickel plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text