Thermo Scientific Chemicals

Calcium bromide hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 018820.22
100 g, Each
Thermo Scientific Chemicals

Calcium bromide hydrate, 95%, Thermo Scientific Chemicals

Catalog number: 018820.22
100 g, Each
Quantity
Catalog number: 018820.22
also known as 018820-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71626-99-8
IUPAC Name
calcium dibromide
Molecular Formula
Br2Ca
InChI Key
WGEFECGEFUFIQW-UHFFFAOYSA-L
SMILES
[Ca++].[Br-].[Br-]
Assay (unspecified)
95% CaBr₂ min

Description

Calcium bromide hydrate is used as freezing mixtures, food preservatives and photography. Its aqueous solution is used in drilling fluids and fire retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text