Calcium bromide hydrate, 95%, Thermo Scientific Chemicals
Calcium bromide hydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium bromide hydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 018820.36
also known as 018820-36
Price (USD)/ Each
97.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
97.00
Add to cart
Calcium bromide hydrate, 95%, Thermo Scientific Chemicals
Catalog number018820.36
Price (USD)/ Each
97.00
-
Add to cart
Chemical Identifiers
CAS71626-99-8
IUPAC Namecalcium dibromide
Molecular FormulaBr2Ca
InChI KeyWGEFECGEFUFIQW-UHFFFAOYSA-L
SMILES[Ca++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95% CaBr₂ min
Calcium bromide hydrate is used as freezing mixtures, food preservatives and photography. Its aqueous solution is used in drilling fluids and fire retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only