Thermo Scientific Chemicals

Selenic acid, 40% aq. soln., Thermo Scientific Chemicals

Catalog number: 018851.22
100 g, Each
Thermo Scientific Chemicals

Selenic acid, 40% aq. soln., Thermo Scientific Chemicals

Catalog number: 018851.22
100 g, Each
Quantity
Catalog number: 018851.22
also known as 018851-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7783-08-6
IUPAC Name
dipotassium selenate
Molecular Formula
K2O4Se
InChI Key
YAZJAPBTUDGMKO-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-][Se]([O-])(=O)=O
Concentration (weight/weight)
40% min

Description

Selenic acid is used as an oxidizing agent. Its derivative sodium selenate is used in the preparation of glass and animal feeds. It acts as an analytical reagent and also used for the preparation of other selenium salts such as gold(III) selenate. Further, it reacts with fluorosulfuric acid to get selenoyl fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute Inhalation Toxicity - Vapors
    Category 3
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H331 - Toxic if swallowed or if inhaled

    H373 - May cause damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P260 - Do not breathe dust/fume/gas/mist/vapors/spray

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P311 - Call a POISON CENTER or doctor/physician

    P403 + P233 - Store in a well-ventilated place. Keep container tightly closed