Thermo Scientific Chemicals

Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals

Catalog number: 018859.09
10 g, Each
Thermo Scientific Chemicals

Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals

Catalog number: 018859.09
10 g, Each
Quantity
Catalog number: 018859.09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18267-08-8
IUPAC Name
zirconium(4+) ethanolate
Molecular Formula
C2H5OZr
InChI Key
VCYRFVPNTBNCLW-UHFFFAOYSA-N
SMILES
[Zr+4].CC[O-]
Appearance (Color)
White to pale cream
Form
Crystalline powder or powder and/or chunks
Assay from Suppliers CofA
≥98.5%

Description

Zirconium(IV) ethoxide is used as precursor for the synthesis of olefin-functionalized zirconium-containing monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text