Thermo Scientific™

Calcium hydride, 90-95%, Thermo Scientific Chemicals

Catalog number: 019106.22
100 g, Each
Thermo Scientific™

Calcium hydride, 90-95%, Thermo Scientific Chemicals

Catalog number: 019106.22
100 g, Each
Quantity
Catalog number: 019106.22
also known as 019106-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-78-8
IUPAC Name
calcium
Molecular Formula
Ca
InChI Key
OYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES
[Ca]
Appearance (Color)
White to gray
Form
Granules or powder or granular powder
Assay from Supplier's CofA
≥88.0%

Description

Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text