Copper(I) bromide-dimethyl sulfide complex, Thermo Scientific Chemicals
Copper(I) bromide-dimethyl sulfide complex, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(I) bromide-dimethyl sulfide complex, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number 019368.18
also known as 019368-18
Price (USD)/ Each
260.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
260.00
Add to cart
Copper(I) bromide-dimethyl sulfide complex, Thermo Scientific Chemicals
Catalog number019368.18
Price (USD)/ Each
260.00
-
Add to cart
Chemical Identifiers
CAS54678-23-8
IUPAC Name(methylsulfanyl)methane; bromocopper
Molecular FormulaC2H6BrCuS
InChI KeyPMHQVHHXPFUNSP-UHFFFAOYSA-M
SMILES[Cu]Br.CSC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Iodometric Titration)> 97.0%
Appearance (Color)White to off-white to faint green
Formpowder
Copper(I) bromide-dimethyl sulfide complex is used as catalyst for adding Grignard reagents to fullerenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only