Thermo Scientific Chemicals

Pyridine, ACS, 99.0+%, Thermo Scientific Chemicals

Catalog number: 019378.K7
4 L, Each
Thermo Scientific Chemicals

Pyridine, ACS, 99.0+%, Thermo Scientific Chemicals

Catalog number: 019378.K7
4 L, Each
Quantity
Catalog number: 019378.K7
also known as 019378-K7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
110-86-1
IUPAC Name
pyridine
Molecular Formula
C5H5N
InChI Key
JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES
C1=CC=NC=C1
Color (APHA)
≤10 ( at time of packaging)
Water Content (Karl Fischer Titration)
≤0.1%
Residue on Evaporation
≤0.002%
Appearance (Color)
Clear, colorless to pale yellow
Comment
Permanganate reducing substances: Passes Test

Description

As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane (C5H5NBH3) and pyridine-sulfur trioxide (C5H5NSO3) are widely used owing to their superior characteristics in their reactivity and solubility characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text