1,2-Dichlorobenzene, HPLC Grade, 98% min, Thermo Scientific Chemicals
1,2-Dichlorobenzene, HPLC Grade, 98% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dichlorobenzene, HPLC Grade, 98% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
4 L
Catalog number 019385.K2
also known as 019385-K2
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
Add to cart
1,2-Dichlorobenzene, HPLC Grade, 98% min, Thermo Scientific Chemicals
Catalog number019385.K2
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS95-50-1
IUPAC Name1,2-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyRFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at400nm: 0.01 AU, max
Residue on Evaporation5ppm
FormLiquid
Assay (GC)≥98.0%
Water Content (Karl Fischer Titration)0.02%, max
View more
1,2-Dichlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only