Thermo Scientific Chemicals

1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 019390.K7
4 L, Each
Thermo Scientific Chemicals

1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 019390.K7
4 L, Each
Quantity
Catalog number: 019390.K7
also known as 019390-K7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120-82-1
IUPAC Name
1,2,4-trichlorobenzene
Molecular Formula
C6H3Cl3
InChI Key
PBKONEOXTCPAFI-UHFFFAOYSA-N
SMILES
ClC1=CC=C(Cl)C(Cl)=C1
Form
Liquid
Comment
Country of origin:
Optical Absorbance at
310nm: ≤0.5 AU
Residue on Evaporation
≤5ppm
Assay (GC)
≥99.0%

Description

1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text