Methanol, Semiconductor Grade, 99.9% min, Thermo Scientific Chemicals
Methanol, Semiconductor Grade, 99.9% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol, Semiconductor Grade, 99.9% min, Thermo Scientific Chemicals

Maximum level of impurities: Color (APHA) 10, Acidity 0.3meq/g, Alkalinity 0.1meq/g, Evaporation residue 5ppm, H2O (by Karl Fischer titrn) 0.05%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.5ppm
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
4 x 1 L
Catalog number 019393.K7
also known as 019393-K7
Price (USD)/ Each
180.00
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
180.00
Add to cart
Methanol, Semiconductor Grade, 99.9% min, Thermo Scientific Chemicals
Catalog number019393.K7
Price (USD)/ Each
180.00
-
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water(by Karl Fischer titrn) 0.05%
Residue on Evaporation5ppm
Color (APHA)10
Assay (GC)99.9% min
Free acid (titration)Acidity 0.3meq/g
View more
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only