2-Propanol, Semiconductor Grade, 99.5% min, Thermo Scientific Chemicals
2-Propanol, Semiconductor Grade, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Propanol, Semiconductor Grade, 99.5% min, Thermo Scientific Chemicals

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Solubility in H2O P.T., H2O (by Karl Fischer titrn) 0.05%, Acidity 0.2meq/g, Alkalinity 0.1meq/g, Cl 0.2ppm, PO4 0.5ppm
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 x 1 L
4 L
4 x 4 L
Catalog number 019397.AP
also known as 019397-AP
Price (USD)/ Each
42.65
Online exclusive
47.50 
Save 4.85 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
47.50 
Save 4.85 (10%)
Add to cart
2-Propanol, Semiconductor Grade, 99.5% min, Thermo Scientific Chemicals
Catalog number019397.AP
Price (USD)/ Each
42.65
Online exclusive
47.50 
Save 4.85 (10%)
-
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentPO₄ 0.5ppm, max
Water Content (Karl Fischer Titration)0.05%, max
Residue on Evaporation5ppm, max
Solution Testin H₂O P.T.
FormClear liquid
View more
2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only