Thermo Scientific Chemicals

Trichloroethylene, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 019401.K7
4 L, Each
Thermo Scientific Chemicals

Trichloroethylene, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 019401.K7
4 L, Each
Quantity
Catalog number: 019401.K7
also known as 019401-K7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-01-6
IUPAC Name
1,1,2-trichloroethene
Molecular Formula
C2HCl3
InChI Key
XSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILES
ClC=C(Cl)Cl
Heavy Metals (as Pb)
≤1ppm
Residue on Evaporation
≤0.001%
Stabilizer
If present: Report amount and type of stabilizer
Color (APHA)
≤10
Appearance (Color)
Clear colorless

Description

Trichloroethylene is used as solvent for paints, dyes, waxes, oils, fats and resins, as an additive in polymerization reactions, as a reactant to prepare various halohydrocarbons (1,1,1,2-tetrafluoroethane) and other chemicals, as a coolant for cutting tantalum and acts as good heat transfer medium. It is used in fire extinguisher fluids and in the recovery of oils and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text