Thermo Scientific Chemicals

Zinc iron oxide, Thermo Scientific Chemicals

Catalog number: 019407.14
25 g, Each
Thermo Scientific Chemicals

Zinc iron oxide, Thermo Scientific Chemicals

Catalog number: 019407.14
25 g, Each
Quantity
Catalog number: 019407.14
also known as 019407-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12063-19-3
IUPAC Name
bis(ironoylol) zinc
Molecular Formula
Fe2H2O4Zn
InChI Key
NNGHIEIYUJKFQS-UHFFFAOYSA-L
SMILES
[Zn].O[Fe]=O.O[Fe]=O
Impurity content
1% max

Description

Zinc iron oxide is used as pigments, especially in applications related to heat stability. It is also used in high corrosion-resistant coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text