1x1 image pixel for data collection
Nickel chromite
Thermo Scientific Chemicals
Nickel chromite
CAS: 12018-18-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 019424.14
also known as 019424-14
Price (USD)
99.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
99.00
Each
Add to cart
Chemical Identifiers
CAS12018-18-7
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max.
In mixing Nickel (II) Chromate with Aluminates or Cobalt Chromate, a binary compound known as a spinel is formed that is able to withstand temperatures up to 1000 °C. The products from these mixings can be used as oxidation-resistive components in airplane and space shuttle coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection