1-Cyclohexene-1-acetic acid, Thermo Scientific Chemicals
1-Cyclohexene-1-acetic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Cyclohexene-1-acetic acid, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 019462.06
also known as 019462-06
Price (USD)/ Each
112.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
112.00
Add to cart
1-Cyclohexene-1-acetic acid, Thermo Scientific Chemicals
Catalog number019462.06
Price (USD)/ Each
112.00
-
Add to cart
Specifications
Chemical Name or Material1-Cyclohexene-1-acetic acid
CAS18294-87-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
1-Cyclohexene-1-acetic acid used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only