Thermo Scientific Chemicals

Tetraethylene glycol dimethyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: 020443.A1
1 kg, Each
Thermo Scientific Chemicals

Tetraethylene glycol dimethyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: 020443.A1
1 kg, Each
Quantity
Catalog number: 020443.A1
also known as 020443-A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
143-24-8
IUPAC Name
2,5,8,11,14-pentaoxapentadecane
Molecular Formula
C10H22O5
InChI Key
ZUHZGEOKBKGPSW-UHFFFAOYSA-N
SMILES
COCCOCCOCCOCCOC
Appearance (Color)
Clear, colorless
Form
Liquid
Assay (GC)
>98.0%

Description

Tetraethylene glycol dimethyl ether is used in lithium-ion battery technology. It is associated with trifluoroethanol and used as a working pair for organic absorption heat pumps. It acts as a solvent for cleaning and degreasing due to its chemical and thermal stability. Moreover, it is suitable solvent for high temperature reactions due its high boiling point. Further, it is used in the selective adsorption of proteins during the promotion of cell adhesion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text