Thermo Scientific Chemicals

n-Octadecylphosphonic acid, 97%, Thermo Scientific Chemicals

Catalog number: 020645.03
1 g, Each
Thermo Scientific Chemicals

n-Octadecylphosphonic acid, 97%, Thermo Scientific Chemicals

Catalog number: 020645.03
1 g, Each
Quantity
Catalog number: 020645.03
also known as 020645-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4724-47-4
IUPAC Name
octadecylphosphonic acid
Molecular Formula
C18H39O3P
InChI Key
FTMKAMVLFVRZQX-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCP(O)(O)=O
Appearance (Color)
White to pale cream
Assay (GC)
≥96.0%
Melting Point (DSC)
95 deg C, min

Description

n-Octadecylphosphonic acid is used in thermal paper for receipts, adding machines and tickets. It acts as surfactant, emulsifier, dispersants and chelating agents. Further, it is a long chain phosphonic acid compound utilized for proteomics research. In addition to this, it is used to prepare modified mesoporous magnetic nano particles through Lewis acid/base interaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text