Thermo Scientific Chemicals

1-Octylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: 020651.03
1 g, Each
Thermo Scientific Chemicals

1-Octylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog number: 020651.03
1 g, Each
Quantity
Catalog number: 020651.03
also known as 020651-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4724-48-5
IUPAC Name
octylphosphonic acid
Molecular Formula
C8H19O3P
InChI Key
NJGCRMAPOWGWMW-UHFFFAOYSA-N
SMILES
CCCCCCCCP(O)(O)=O
Assay (GC)
> 97.5%
Form
powder
Appearance (Color)
White to pale cream

Description

1-Octylphosphonic acid is a basic building block and used in the preparation of various pharmaceutical compounds and chemical products. It is supported with calcium hydroxyapatite and used in the dental field as corrosion inhibitor and organic building block. It acts as a corrosion inhibitor in metallic household surfaces such as faucets and drains as well as prevents the rusting of can or dispenser. It is also used in the synthesis of octyl-phosphonic acid dimethyl ester by reacting with iodomethane in presence of CS2 CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text