2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 020748.06
also known as 020748-06
Price (USD)/ Each
314.65
Online exclusive
349.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
314.65
Online exclusive
349.00 
Save 34.35 (10%)
Add to cart
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
Catalog number020748.06
Price (USD)/ Each
314.65
Online exclusive
349.00 
Save 34.35 (10%)
-
Add to cart
Specifications
Chemical Name or Material2,6-Dimethylphenyl isocyanide
Melting Point73°C to 74°C
CAS2769-71-3
Health Hazard 1H301+H311-H330
Health Hazard 2GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
View more
2,6-Dimethylphenyl isocyanate is used in the preparation of homoleptic metallaamidine complex, derivatized bets-cyclodextrins. It is used as chiral stationary phase in normal-phase liquid chromatography. It is also involved in the preparation of tris( 2,6-dimethylphenylimido)methylrhenium (VII) and 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only