Thermo Scientific Chemicals

Sodium trifluoromethanesulfonate, Thermo Scientific Chemicals

Catalog number: 020837.09
10 g, Each
Thermo Scientific Chemicals

Sodium trifluoromethanesulfonate, Thermo Scientific Chemicals

Catalog number: 020837.09
10 g, Each
Quantity
Catalog number: 020837.09
also known as 020837-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2926-30-9
IUPAC Name
sodium trifluoromethanesulfonate
Molecular Formula
CF3NaO3S
InChI Key
XGPOMXSYOKFBHS-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)C(F)(F)F
Purity
≥95.0%
Assay (unspecified)
Sodium (Na): ≥12.7%
Form
Powder

Description

Catalyzes the aza Diels-Alder reaction of Danishefsky's diene with imines in water under neutral conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text