Thermo Scientific Chemicals

Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 020932.LV
1 gal., Each
Thermo Scientific Chemicals

Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 020932.LV
1 gal., Each
Quantity
Catalog number: 020932.LV
also known as 020932-LV
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-59-2
IUPAC Name
tetramethylazanium hydroxide
Molecular Formula
C4H13NO
InChI Key
WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES
[OH-].C[N+](C)(C)C
Form
Liquid
Color (APHA)
≤10
Assay (Aqueous acid-base Titration)
24.90-25.10 wt%
Carbonate
≤300 ppm
Ion Chromatography
≤5 ppm Halide ( as Chloride)

Description

Tetramethylammonium hydroxide is used to produce tetramethyl-ammonium azide. It is used as an anisotropic etchant of silicon, basic solvent in the development of acidic photoresist in photolithography process, as surfactant in the synthesis of ferrofluid and as a polarographic reagent. It finds application in the production of organic silicon, where it is used for computer silicon wafer surface brightener and cleaning agent. It is also involved in the purification of some metallic elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text