Thermo Scientific Chemicals

Cerium(III) fluoride, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 021118.36
500 g, Each
Thermo Scientific Chemicals

Cerium(III) fluoride, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 021118.36
500 g, Each
Quantity
Catalog number: 021118.36
also known as 021118-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-88-5
IUPAC Name
cerium(3+) trifluoride
Molecular Formula
CeF3
InChI Key
QCCDYNYSHILRDG-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Ce+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Cerium(III) fluoride is used to make Lewis acid. It is considered to be the most effective glass polishing agent for high precision optical polishing. Cerium fluoride used as a core additive to improve the properties of the eventual weld. The fluoride provides stability to the arc and also modifies the behavior of the molten metal flowing into the weld.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text