Thermo Scientific Chemicals

Cerium(IV) oxide, hydrated, Thermo Scientific Chemicals

Catalog number: 021120.18
50 g, Each
Thermo Scientific Chemicals

Cerium(IV) oxide, hydrated, Thermo Scientific Chemicals

Catalog number: 021120.18
50 g, Each
Quantity
Catalog number: 021120.18
also known as 021120-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12014-56-1
Molecular Formula
CeH8O4
InChI Key
DPUCLPLBKVSJIB-UHFFFAOYSA-N
SMILES
O.O.O.O.[Ce]
Molecular Weight (g/mol)
212.18
Insoluble material
0.25% max
Impurity content
0.1% PO₄ max
Impurity content
0.1% Fe max

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text