Cerium(IV) oxide, hydrated, Thermo Scientific Chemicals
Cerium(IV) oxide, hydrated, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(IV) oxide, hydrated, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 021120.30
also known as 021120-30
Price (USD)/ Each
198.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
198.00
Add to cart
Cerium(IV) oxide, hydrated, Thermo Scientific Chemicals
Catalog number021120.30
Price (USD)/ Each
198.00
-
Add to cart
Chemical Identifiers
CAS12014-56-1
Molecular FormulaCeH8O4
InChI KeyDPUCLPLBKVSJIB-UHFFFAOYSA-N
SMILESO.O.O.O.[Ce]
Molecular Weight (g/mol)212.18
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.25% max
Impurity content0.1% PO₄ max
Impurity content0.1% Fe max
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only