Thermo Scientific Chemicals

Cerium(III) oxalate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 021122.22
100 g, Each
Thermo Scientific Chemicals

Cerium(III) oxalate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 021122.22
100 g, Each
Quantity
Catalog number: 021122.22
also known as 021122-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15750-47-7
IUPAC Name
dicerium(3+) trioxalate
Molecular Formula
C6Ce2O12
InChI Key
ZMZNLKYXLARXFY-UHFFFAOYSA-H
SMILES
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text