Thermo Scientific Chemicals

Calcium peroxide, typically 65%, Thermo Scientific Chemicals

Catalog number: 021157.22
100 g, Each
Thermo Scientific Chemicals

Calcium peroxide, typically 65%, Thermo Scientific Chemicals

Catalog number: 021157.22
100 g, Each
Quantity
Catalog number: 021157.22
also known as 021157-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1305-79-9
IUPAC Name
calcium dioxidanediide
Molecular Formula
CaO2
InChI Key
LHJQIRIGXXHNLA-UHFFFAOYSA-N
SMILES
[Ca++].[O-][O-]
Assay (unspecified)
65% min

Description

Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text