Potassium chloride, Optical Grade, Thermo Scientific Chemicals
Potassium chloride, Optical Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium chloride, Optical Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 022109.22
also known as 022109-22
Price (USD)/ Each
183.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
183.00
Add to cart
Potassium chloride, Optical Grade, Thermo Scientific Chemicals
Catalog number022109.22
Price (USD)/ Each
183.00
-
Add to cart
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentTypical transmission range 0.21 - 20 microns
FormBroken crystals
Potassium chloride is a salt commonly used in the preparation of phosphate buffered saline and for the extraction and solubilization of proteins. It is a good potassium source. Also used in the hypotonic KCl-propidium iodide protocol for univariate analysis of metaphase chromosomes ; dissociation of eukaryotic ribosomes into subunits by high concentrations of potassium (0.3-1.0 M); preparation of mitochondria, provides an ionic osmotic support for those tissues that assume a gelatinous consistency upon homogenization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only