Lithium chloride monohydrate, 99.95% (metals basis), Thermo Scientific Chemicals
Lithium chloride monohydrate, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium chloride monohydrate, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 022151.14
also known as 022151-14
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
Add to cart
Lithium chloride monohydrate, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number022151.14
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Chemical Identifiers
CAS16712-20-2
IUPAC Namelithium(1+) hydrate chloride
Molecular FormulaClH2LiO
InChI KeyVXJIMUZIBHBWBV-UHFFFAOYSA-M
SMILES[Li+].O.[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Lithium chloride monohydrate is used as a precursor in the production of lithium metal and other lithium compounds. It acts as a welding agent for aluminum, salt bath for heat -treatment by low temperature and soldering techniques. It is also used as a tracer for chemical products viz. denaturation of wine. For absorbers, it is an absorption and disinfection reagent as well as desiccant for air conditioner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only