Trichloroacetic acid, ACS, 99% min, Thermo Scientific Chemicals
Trichloroacetic acid, ACS, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroacetic acid, ACS, 99% min, Thermo Scientific Chemicals

Protein precipitation reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 022156.A1
also known as 022156-A1
Price (USD)/ Each
315.65
Online exclusive
351.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
315.65
Online exclusive
351.00 
Save 35.35 (10%)
Add to cart
Trichloroacetic acid, ACS, 99% min, Thermo Scientific Chemicals
Catalog number022156.A1
Price (USD)/ Each
315.65
Online exclusive
351.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS76-03-9
IUPAC Nametrichloroacetic acid
Molecular FormulaC2HCl3O2
InChI KeyYNJBWRMUSHSURL-UHFFFAOYSA-N
SMILESOC(=O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.03%
FormLiquid
Impurity contentChlorine (Cl): 0.002%
Impurity contentNitrate (NO3): 0.002%
Impurity contentSubstances darkened by sulfuric acid P.T.
View more
Protein precipitation reagentTrichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used in cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and as a decalcifier and fixative in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only