1,3-Dichloroacetone, typically 99%, Thermo Scientific Chemicals
1,3-Dichloroacetone, typically 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dichloroacetone, typically 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number 022302.22
also known as 022302-22
Price (USD)/ Each
269.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
269.00
Add to cart
1,3-Dichloroacetone, typically 99%, Thermo Scientific Chemicals
Catalog number022302.22
Price (USD)/ Each
269.00
-
Add to cart
Specifications
Chemical Name or Material1,3-Dichloroacetone
CAS534-07-6
Health Hazard 1H300+H310+H330-H314-H335-H340
Health Hazard 2GHS H Statement
H300-H310-H330-H314-H318-H341
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Health Hazard 3P201-P202-P260-P262-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
1,3-Dichloroacetone was used as acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanon

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only