Samarium(II) iodide, 0.07 - 0.12M in THF, stab., Thermo Scientific Chemicals
Samarium(II) iodide, 0.07 - 0.12M in THF, stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium(II) iodide, 0.07 - 0.12M in THF, stab., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 022360.AE
also known as 022360-AE
Price (USD)/ Each
92.80
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
92.80
Add to cart
Samarium(II) iodide, 0.07 - 0.12M in THF, stab., Thermo Scientific Chemicals
Catalog number022360.AE
Price (USD)/ Each
92.80
-
Add to cart
Chemical Identifiers
CAS32248-43-4
IUPAC Nameλ²-samarium(2+) diiodide
Molecular FormulaI2Sm
InChI KeyUAWABSHMGXMCRK-UHFFFAOYSA-L
SMILES[I-].[I-].[Sm++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to very dark blue
Concentration≥0.075M
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only