Thermo Scientific Chemicals

Tin(II) acetate, 95%, Thermo Scientific Chemicals

Catalog number: 022361.06
5 g, Each
Thermo Scientific Chemicals

Tin(II) acetate, 95%, Thermo Scientific Chemicals

Catalog number: 022361.06
5 g, Each
Quantity
Catalog number: 022361.06
also known as 022361-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
638-39-1
Assay (unspecified)
94.1% min

Description

Tin(II) acetate is used as a precursor in the production of tin anode, Sn-Cu bimetallic nanoparticles, high surface area tin oxide catalysts. It acts as Lewis acid catalysts for the esterification and transesterification reactions. It serves as an efficient catalytic system for carbon-hydrogen activation of methoxy group of anisole in presence of palladium(II) acetate. It plays an important role in stannous oral care compositions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text