Sodium tetrachloroaluminate, Thermo Scientific Chemicals
Sodium tetrachloroaluminate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tetrachloroaluminate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 022375.14
also known as 022375-14
Price (USD)/ Each
292.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
292.00
Add to cart
Sodium tetrachloroaluminate, Thermo Scientific Chemicals
Catalog number022375.14
Price (USD)/ Each
292.00
-
Add to cart
Chemical Identifiers
CAS7784-16-9
IUPAC Namesodium tetrachloroalumanuide
Molecular FormulaAlCl4Na
InChI KeyCMLRNXGMTZKKSR-UHFFFAOYSA-J
SMILES[Na+].Cl[Al-](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Cl 72% min
Sodium tetrachloroaluminate is used as Friedel-Crafts acylating agent , dehydrating and dehydrogenating agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only