Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid tripotassium salt, Thermo Scientific Chemicals

Catalog number: 022425.22
100 g, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid tripotassium salt, Thermo Scientific Chemicals

Catalog number: 022425.22
100 g, Each
Quantity
Catalog number: 022425.22
also known as 022425-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17572-97-3
Appearance (Color)
White
Form
Crystalline powder
Assay (unspecified)
> 98.0%

Description

Ethylenediaminetetraacetic acid tripotassium salt is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. And it is also used to study chelators, biochemicals and reagents. EDTA tripotassium salt has been used in a study to assess the safety of EDTA and HEDTA ingredients in cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text