Chemical Identifiers
CAS14025-21-9
IUPAC Namezinc(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular FormulaC10H12N2Na2O8Zn
InChI KeyUBLOJEHIINPTTG-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View more