Thermo Scientific Chemicals

Strontium dodecairon nonadecaoxide, 99.0% (metals basis), Thermo Scientific Chemicals

Catalog number: 022569.06
5 g, Each
Thermo Scientific Chemicals

Strontium dodecairon nonadecaoxide, 99.0% (metals basis), Thermo Scientific Chemicals

Catalog number: 022569.06
5 g, Each
Quantity
Catalog number: 022569.06
also known as 022569-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12023-91-5
IUPAC Name
diiron(3+) distrontium(2+) pentaoxidandiide
Molecular Formula
Fe2O5Sr2
InChI Key
GKHGYOYJUWWDJG-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[O--].[Fe+3].[Fe+3].[Sr++].[Sr++]
Appearance (Color)
Dark purple
Form
Powder
Total Metal Impurities
1.0% max.

Description

Strontium dodecairon nonadecaoxide is used in manufacture of mixtures, granulates for plastic articles, production of magnet articles by compacting, sintering or other processes. And it is also used as an additive to flares, pyrotechnics, provides high refractive index to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text