Strontium dodecairon nonadecaoxide, 99.0% (metals basis), Thermo Scientific Chemicals
Strontium dodecairon nonadecaoxide, 99.0% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium dodecairon nonadecaoxide, 99.0% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 022569.14
also known as 022569-14
Price (USD)/ Each
336.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
336.00
Add to cart
Strontium dodecairon nonadecaoxide, 99.0% (metals basis), Thermo Scientific Chemicals
Catalog number022569.14
Price (USD)/ Each
336.00
-
Add to cart
Chemical Identifiers
CAS12023-91-5
IUPAC Namediiron(3+) distrontium(2+) pentaoxidandiide
Molecular FormulaFe2O5Sr2
InChI KeyGKHGYOYJUWWDJG-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[O--].[Fe+3].[Fe+3].[Sr++].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark purple
FormPowder
Total Metal Impurities1.0% max.
Strontium dodecairon nonadecaoxide is used in manufacture of mixtures, granulates for plastic articles, production of magnet articles by compacting, sintering or other processes. And it is also used as an additive to flares, pyrotechnics, provides high refractive index to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only