Thermo Scientific Chemicals

Ethanethiol, 97%, Thermo Scientific Chemicals

Catalog number: 022585.AP
500 mL, Each
Thermo Scientific Chemicals

Ethanethiol, 97%, Thermo Scientific Chemicals

Catalog number: 022585.AP
500 mL, Each
Quantity
Catalog number: 022585.AP
also known as 022585-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-08-1
IUPAC Name
ethanethiol
Molecular Formula
C2H6S
InChI Key
DNJIEGIFACGWOD-UHFFFAOYSA-N
SMILES
CCS
Appearance (Color)
Colorless
Form
Liquid
Assay (GC)
≥96.0%

Description

Ethanethiol acts as a stabilizer for adhesives and an odorant for natural gas. It is used in the odorless liquefied petroleum gas (LPG) in order to find out the gas leaks. It is used as key starting material for the preparation of ethylsulfonic acid, diethyldisulfide and sodium ethylmercaptide. Further, it is also used in the underground mining industry as stench gas. In organic synthesis, it gives powerful nucleophile set- in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text