Barium aluminum oxide, tech., Thermo Scientific Chemicals
Barium aluminum oxide, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium aluminum oxide, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 022642.36
also known as 022642-36
Price (USD)/ Each
229.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
229.00
Add to cart
Barium aluminum oxide, tech., Thermo Scientific Chemicals
Catalog number022642.36
Price (USD)/ Each
229.00
-
Add to cart
Chemical Identifiers
CAS12004-04-5
IUPAC Namedialuminium(3+) dibarium(2+) pentaoxidandiide
Molecular FormulaAl2Ba2O5
InChI KeyWIHSDLARTQXPRC-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Ba++].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ba 47% min
Barium aluminum oxide is used for getter alloys in vacuum tubes. It also finds application as deoxidizer for copper, Frary’s metal, lubricant for anode rotors in x-ray tubes and spark-plug alloys. It is used to prepare heat-resistant refractory and super-refractory concretes. Further, it gives a better protection against gamma radiation and X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only