Thermo Scientific Chemicals

Calcium aluminum oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 022646.06
5 g, Each
Thermo Scientific Chemicals

Calcium aluminum oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 022646.06
5 g, Each
Quantity
Catalog number: 022646.06
also known as 022646-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12042-68-1
IUPAC Name
calcium dialumanoylolate
Molecular Formula
Al2CaO4
InChI Key
XFWJKVMFIVXPKK-UHFFFAOYSA-N
SMILES
[Ca++].[O-][Al]=O.[O-][Al]=O
Total Metal Impurities
1% max.

Description

Calcium aluminum oxide is used as a precursor in the production of naphtha steam reforming catalysts, refinery gas steam reforming catalysts, aluminum chlorohydrate, secondary hydrocarbon steam reforming catalysts. It is also used in the production of refractory and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text