Lanthanum(III) sulfide, 99% (REO), Thermo Scientific Chemicals
Lanthanum(III) sulfide, 99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) sulfide, 99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 022655.04
also known as 022655-04
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
Add to cart
Lanthanum(III) sulfide, 99% (REO), Thermo Scientific Chemicals
Catalog number022655.04
Price (USD)/ Each
108.65
Online exclusive
120.00 
Save 11.35 (9%)
-
Add to cart
Chemical Identifiers
CAS12031-49-1
IUPAC Namedilanthanum(3+) trisulfanediide
Molecular FormulaLa2S3
InChI KeyYTYSNXOWNOTGMY-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[La+3].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities1% max.
Lanthanum(III) sulfide is used as a precursor in the production of gallium lanthanum sulfide and strontium lathanum sulfide. It is also used in glass manufacturing. It is doped with other metals and used in optical fibers. It is also used as optical switching, high power-delivery, acousto-optic devices, air-guiding fibers, fiber sensors and mid-IR devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only