Thermo Scientific Chemicals

Silver(II) oxide, 98%, Thermo Scientific Chemicals

Catalog number: 022695.06
5 g, Each
Thermo Scientific Chemicals

Silver(II) oxide, 98%, Thermo Scientific Chemicals

Catalog number: 022695.06
5 g, Each
Quantity
Catalog number: 022695.06
also known as 022695-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1301-96-8
IUPAC Name
oxosilver
Molecular Formula
AgO
InChI Key
OTCVAHKKMMUFAY-UHFFFAOYSA-N
SMILES
O=[Ag]
Assay (unspecified)
97.6% min

Description

Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text